N-(Diphenylmethylene)aminoacetonitrile, 99%
N-(Diphenylmethylene)aminoacetonitrile, 99%
N-(Diphenylmethylene)aminoacetonitrile, 99%
Thermo Scientific Chemicals

N-(Diphenylmethylene)aminoacetonitrile, 99%

CAS: 70591-20-7 | C15H12N2 | 220.28 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 124140250
Price (USD)
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS70591-20-7
IUPAC Name2-[(diphenylmethylidene)amino]acetonitrile
Molecular FormulaC15H12N2
InChI KeyVRLJFRODHVSTIK-UHFFFAOYSA-N
SMILESN#CCN=C(C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow to light grey
Melting point83°C to 86°C
Infrared spectrumConforms
HPLC>=98.5 %
Appearance (Form)Powder or crystals
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only