Hydrochloric acid, 1N standard solution
Hydrochloric acid, 1N standard solution
Hydrochloric acid, 1N standard solution
Thermo Scientific Chemicals

Hydrochloric acid, 1N standard solution

CAS: 7647-01-0 | ClH | 36.46 g/mol
数量:
2.5 L
10 L
パッケージ:
すべて表示
Plastic Bottle
製品番号(カタログ番号) 124210025
価格(JPY)
-
数量:
2.5 L
パッケージ:
Plastic Bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS7647-01-0
IUPAC Namehydrogen chloride
Molecular FormulaClH
InChI KeyVEXZGXHMUGYJMC-UHFFFAOYSA-N
SMILESCl
さらに表示
Appearance (Form)Solution
Appearance (Color)Clear colorless
Strength0.998 to 1.002 N (20°C)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Volumetric standard solution
RUO – Research Use Only