Silver nitrate, 0.1 N standard solution
Silver nitrate, 0.1 N standard solution
Silver nitrate, 0.1 N standard solution
Thermo Scientific Chemicals

Silver nitrate, 0.1 N standard solution

CAS: 7761-88-8 | AgNO3 | 169.87 g/mol
数量:
1 L
5 L
パッケージ:
HDPE Bottle
製品番号(カタログ番号) 124250050
価格(JPY)
-
数量:
5 L
パッケージ:
HDPE Bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS7761-88-8
IUPAC Namesilver(1+) nitrate
Molecular FormulaAgNO3
InChI KeySQGYOTSLMSWVJD-UHFFFAOYSA-N
SMILES[Ag+].[O-][N+]([O-])=O
さらに表示
Appearance (Form)Solution
Strength0.0998 to 0.1002 N (20°C)
Appearance (Color)Clear colorless
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Volumetric standard solution
RUO – Research Use Only