Sodium hydroxide, 1N standard solution
Sodium hydroxide, 1N standard solution
Sodium hydroxide, 1N standard solution
Thermo Scientific Chemicals

Sodium hydroxide, 1N standard solution

CAS: 1310-73-2 | HNaO | 39.997 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
5 L
25 L
Packaging:
View all
Plastic Can
Catalog number 124260250
Price (USD)
352.65
Online Exclusive
392.00
Save 39.35 (10%)
Each
Quantity:
25 L
Packaging:
Plastic Can
Request bulk or custom format
Price (USD)
352.65
Online Exclusive
392.00
Save 39.35 (10%)
Each
Chemical Identifiers
CAS1310-73-2
IUPAC Namesodium hydroxide
Molecular FormulaHNaO
InChI KeyHEMHJVSKTPXQMS-UHFFFAOYSA-M
SMILES[OH-].[Na+]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Strength0.995 to 1.005 N (20°C)
Appearance (Form)Solution
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Volumetric standard solution
RUO – Research Use Only