Interested in a bulk or custom version of this product? Request pricing.

C6 Recombinant Human Protein, His tag

Catalog number: 12426H08H250

 Related applications: Protein Assays and Analysis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Complement component 6 (C6), recombinant human protein is supplied as a lyophilized powder. In general, recombinant proteins can be used as protein stucture analysis and in cell biology research applications.

This recombinant protein was expressed from a DNA sequence encoding the human C6 (AAA59668.1) (Met 1- Ala 934) fused to a polyhistidine tag at the C-terminus.

Activity: This recombinant protein has not been tested.

Formulation: 140 mM NaCl, 2.7 mM KCl, 10 mM Na2HPO4, 1.8 mM KH2PO4, pH7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween80.

Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2 mg/ml or lower. It is recommended that the protein be aliquoted and be used as soon as possible. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.

Expiration Date: Expires one year from date of receipt when stored as instructed.



This protein is manufactured by SINO Biological.

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Endotoxin Level: < 1 EU/µg
Expression: Mammalian
Form: Lyophilized
Gene Aliases: C6
Gene ID: 729
Gene Symbol: C6
Molecular Weight: 104 kDa
Product Line: Sino Biological®
Product Size: 5 x 50 µg
Protein Family: Other Proteins
Protein Form: Full Length, Recombinant
Protein Name: Complement component 6
Protein Tag or Fusion: His Tag
Purification: PAGE
Purity: ≥ 85% by SDS-PAGE
Research Category: Immunology, Inflammation, Microbiology
Species: Human
Tag Position: C-Terminal
Validated Application: Assay Standard

Contents & storage

Includes 5 vials of lyophilized protein. Store at -20 °C.

Documents

Manuals & protocols