beta-Nicotinamide adenine dinucleotide hydrate, 98+%
beta-Nicotinamide adenine dinucleotide hydrate, 98+%
beta-Nicotinamide adenine dinucleotide hydrate, 98+%
Thermo Scientific Chemicals

beta-Nicotinamide adenine dinucleotide hydrate, 98+%

CAS: 53-84-9 | C21H27N7O14P2 | 663.43 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
5 g
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 124532500
Price (EUR)
27,65
Online exclusive
30,00
Save 2,35 (8%)
Each
Quantity:
250 mg
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
27,65
Online exclusive
30,00
Save 2,35 (8%)
Each
Chemical Identifiers
CAS53-84-9
IUPAC Name1-[(2R,3R,4S,5R)-5-{[({[(2R,3S,4R,5R)-5-(6-amino-9H-purin-9-yl)-3,4-dihydroxyoxolan-2-yl]methoxy}(hydroxy)phosphoryl phosphonato)oxy]methyl}-3,4-dihydroxyoxolan-2-yl]-3-carbamoyl-1λ⁵-pyridin-1-ylium
Molecular FormulaC21H27N7O14P2
InChI KeyBAWFJGJZGIEFAR-WIWLTUSXNA-N
SMILESNC(=O)C1=CC=C[N+](=C1)[C@@H]1O[C@H](COP([O-])(=O)OP(O)(=O)OC[C@H]2O[C@H]([C@H](O)[C@@H]2O)N2C=NC3=C(N)N=CN=C23)[C@@H](O)[C@H]1O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow
Infrared spectrumConforms
Molar absorption coefficient>=17000 l/(mol.cm) (Lambda max 260 nm, pH 7.5)
Solubility(1 % in water) Clear colorless
Appearance (Form)Powder
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only