Calcium carbonate, 98+%, pure, heavy powder
Calcium carbonate, 98+%, pure, heavy powder
Calcium carbonate, 98+%, pure, heavy powder
Thermo Scientific Chemicals

Calcium carbonate, 98+%, pure, heavy powder

CAS: 471-34-1 | CCaO3 | 100.09 g/mol
数量:
1 kg
2.5 kg
10 kg
パッケージ:
すべて表示
Plastic drum
製品番号(カタログ番号) 124670100
価格(JPY)
-
数量:
10 kg
パッケージ:
Plastic drum
一括またはカスタム形式をリクエストする
化学物質識別子
CAS471-34-1
IUPAC Namecalcium carbonate
Molecular FormulaCCaO3
InChI KeyVTYYLEPIZMXCLO-UHFFFAOYSA-L
SMILES[Ca++].[O-]C([O-])=O
さらに表示
Appearance (Form)Powder
Titration Complexometric98.0 to 102.0 %
Appearance (Color)White to almost white to light beige
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only