Tris(dioxa-3,6-heptyl)amine, 95%
Tris(dioxa-3,6-heptyl)amine, 95%
Tris(dioxa-3,6-heptyl)amine, 95%
Thermo Scientific Chemicals

Tris(dioxa-3,6-heptyl)amine, 95%

CAS: 70384-51-9 | C15H33NO6 | 323.42 g/mol
数量:
25 mL
100 mL
パッケージ:
Glass bottle
製品番号(カタログ番号) 124851000
価格(JPY)
-
数量:
100 mL
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS70384-51-9
IUPAC Name8-[2-(2-methoxyethoxy)ethyl]-2,5,11,14-tetraoxa-8-azapentadecane
Molecular FormulaC15H33NO6
InChI KeyXGLVDUUYFKXKPL-UHFFFAOYSA-N
SMILESCOCCOCCN(CCOCCOC)CCOCCOC
さらに表示
Titration with HClO4>=94 %
Refractive index1.4466 to 1.4486 (20°C, 589 nm)
Appearance (Color)Clear light yellow to brown
Infrared spectrumConforms
GC>=94 %
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only