Petroleum ether, extra pure, boiling range 40-60°C, Thermo Scientific Chemicals
Petroleum ether, extra pure, boiling range 40-60°C, Thermo Scientific Chemicals
Petroleum ether, extra pure, boiling range 40-60°C, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Petroleum ether, extra pure, boiling range 40-60°C, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
25 L
Packaging:
View all
Metal Drum
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 124860250
Price (USD)
-
Quantity:
25 L
Packaging:
Metal Drum
Request bulk or custom format
Chemical Identifiers
CAS64742-49-0
IUPAC NamePetroleum ether
MDL NumberMFCD00081849
SpecificationsSpecification SheetSpecification Sheet
Color scale=<10 APHA
Density(20°C) 0.64 to 0.66 g/ml
Residue after evaporation=<0.002 %
Appearance (Form)Clear liquid
Water=<0.03 % (K.F.)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only