Get your genes cloned into this vector with GeneArt Gene Synthesis Services
Product Image
Invitrogen™

pcDNA™3.2-DEST Mammalian Expression Vector

The pcDNA™ vectors are designed for high-level, constitutive expression in a variety of mammalian cell lines. The pcDNA3.2/V5-DEST vector offersRead more
Have Questions?
Catalog NumberQuantity
12489019
also known as 12489-019
6 μg
Catalog number 12489019
also known as 12489-019
Price (USD)
960.00
Each
Quantity:
6 μg
Price (USD)
960.00
Each
The pcDNA™ vectors are designed for high-level, constitutive expression in a variety of mammalian cell lines. The pcDNA3.2/V5-DEST vector offers the following key features:

•Cytomegalovirus (CMV) promoter for high-level expression
•attR sites for Gateway™ cloning, enabling recombination with attL-flanked fragments
•C-terminal V5 tag for easy detection
•Neomycin resistance gene for stable selection
•Ampicillin resistance gene and pUC origin for selection and maintenance in E. coli

Gateway™ Cloning
To fit all of your expression needs, Invitrogen offers state-of-the-art Gateway™ destination vectors for expression in E. coli, insect, yeast, or mammalian cells, as well as for production of native protein or N- or C-terminal fusion proteins. All Gateway™ destination vectors have attR sites for recombination with any attL-flanked fragment, regardless of whether it is an entry clone or an Ultimate™ ORF Clone. The following table lists a variety of available destination vectors.

Additional materials required, available separately: Gateway™ entry clone, appropriate Gateway™ LR Clonase™ enzyme mix, and reaction buffer.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Constitutive or Inducible SystemConstitutive
Delivery TypeTransfection
For Use With (Application)Constitutive Expression
Product TypeMammalian Expression Vector
Quantity6 μg
Selection Agent (Eukaryotic)Geneticin™ (G-418)
VectorpDEST, pcDNA
Cloning MethodGateway
Product LineGateway, pcDNA
PromoterCMV
Protein TagV5 Epitope Tag
Unit SizeEach
Contents & Storage
All destination vectors are provided lyophilized and supercoiled.

Frequently asked questions (FAQs)

I performed stable selection but my antibiotic-resistant clones do not express my gene of interest. What could have gone wrong?

Here are possible causes and solutions:

Detection method may not be appropriate or sensitive enough:
- We recommend optimizing the detection protocol or finding more sensitive methods. If the protein is being detected by Coomassie/silver staining, we recommend doing a western blot for increased sensitivity. The presence of endogenous proteins in the lysate may obscure the protein of interest in a Coomassie/silver stain. If available, we recommend using a positive control for the western blot.
- Insufficient number of clones screened: Screen at least 20 clones.
- Inappropriate antibiotic concentration used for stable selection: Make sure the antibiotic kill curve was performed correctly. Since the potency of a given antibiotic depends upon cell type, serum, medium, and culture technique, the dose must be determined each time a stable selection is performed. Even the stable cell lines we offer may be more or less sensitive to the dose we recommend if the medium or serum is significantly different.
- Expression of gene product (even low level) may not be compatible with growth of the cell line: Use an inducible expression system.
- Negative clones may result from preferential linearization at a vector site critical for expression of the gene of interest: Linearize the vector at a site that is not critical for expression, such as within the bacterial resistance marker.

I used a mammalian expression vector but do not get any expression of my protein. Can you help me troubleshoot?

Here are possible causes and solutions:

- Try the control expression that is included in the kit
Possible detection problem:

- Detection of expressed protein may not be possible in a transient transfection, since the transfection efficiency may be too low for detection by methods that assess the entire transfected population. We recommend optimizing the transfection efficiency, doing stable selection, or using methods that permit examination of individual cells. You can also increase the level of expression by changing the promoter or cell type.
- Expression within the cell may be too low for the chosen detection method. We recommend optimizing the detection protocol or finding more sensitive methods. If the protein is being detected by Coomassie/silver staining, we recommend doing a western blot for increased sensitivity. The presence of endogenous proteins in the lysate may obscure the protein of interest in a Coomassie/silver stain. If available, we recommend using a positive control for the western blot. Protein might be degraded or truncated: Check on a Northern. Possible time-course issue: Since the expression of a protein over time will depend upon the nature of the protein, we always recommend doing a time course for expression. A pilot time-course assay will help to determine the optimal window for expression. Possible cloning issues: Verify clones by restriction digestion and/or sequencing.

Find additional tips, troubleshooting help, and resources within our Protein Expression Support Center.

I am using a mammalian expression vector that has the neomycin resistance gene. Can I use neomycin for stable selection in mammalian cells?

No; neomycin is toxic to mammalian cells. We recommend using Geneticin (a.k.a. G418 Sulfate), as it is a less toxic and very effective alternative for selection in mammalian cells.

Is it okay if my construct has an ATG that is upstream of the ATG in my gene of interest? Will it interfere with translation of my gene?

Translation initiation will occur at the first ATG encountered by the ribosome, although in the absence of a Kozak sequence, initiation will be relatively weak. Any insert downstream would express a fusion protein if it is in frame with this initial ATG, but levels of expressed protein are predicted to be low if there is a non-Kozak consensus sequence. If the vector contains a non-Kozak consensus ATG, we recommend that you clone your gene upstream of that ATG and include a Kozak sequence for optimal expression.

Do you offer a GFP-expressing mammalian expression vector that I can use as a control to monitor my transfection and expression?

We offer pJTI R4 Exp CMV EmGFP pA Vector, Cat. No. A14146, which you can use to monitor your transfection and expression.

Citations & References (2)

Citations & References
Abstract
Endogenous Msx1 antisense transcript: in vivo and in vitro evidences, structure, and potential involvement in skeleton development in mammals.
Authors: Blin-Wakkach C; Lezot F; Ghoul-Mazgar S; Hotton D; Monteiro S; Teillaud C; Pibouin L; Orestes-Cardoso S; Papagerakis P; Macdougall M; Robert B; Berdal A;
Journal:Proc Natl Acad Sci U S A
PubMed ID:11390985
'Msx1 is a key factor for the development of tooth and craniofacial skeleton and has been proposed to play a pivotal role in terminal cell differentiation. In this paper, we demonstrated the presence of an endogenous Msx1 antisense RNA (Msx1-AS RNA) in mice, rats, and humans. In situ analysis revealed ... More
Serinc, an activity-regulated protein family, incorporates serine into membrane lipid synthesis.
Authors:Inuzuka M, Hayakawa M, Ingi T,
Journal:J Biol Chem
PubMed ID:16120614
Cell membranes contain various transporter proteins, some of which are responsible for transferring amino acids across membrane. In this study, we report another class of carrier proteins, termed Serinc1-5, that incorporates a polar amino acid serine into membranes and facilitates the synthesis of two serine-derived lipids, phosphatidylserine and sphingolipids. Serinc ... More