CD CHO AGT™ Medium
Gibco™

CD CHO AGT™ Medium

CD CHO AGT™ (Advanced Granulation Technology) medium is a protein-free, chemically-defined medium optimized for the growth of Chinese hamster ovaryRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
12490001100 L
124900171 L
12490025
also known as 12490-025
10 L
Catalog number 12490001
Price (USD)
7,640.00
Each
Quantity:
100 L
Customize this product
Price (USD)
7,640.00
Each
CD CHO AGT™ (Advanced Granulation Technology) medium is a protein-free, chemically-defined medium optimized for the growth of Chinese hamster ovary (CHO) cells and expression of recombinant proteins in suspension culture. The AGT™ dry media format is designed to provide increased consistency and productivity across all stages of production from development to commercial manufacturing. CD CHO AGT™ medium contains no proteins or peptide components of animal, plant, or synthetic origin, as well as no undefined lysates or hydrolysates.

When to Use CD CHO Medium
• Your GS CHO cells are recommended to be grown in CD CHO medium
• Your cells have been successfully cultured in CD CHO medium in the past and your process is nearing a regulatory filing (DMF available)

When to Consider Another Gibco™ Medium

Consider CD FortiCHO™ medium when:
• The cell line you are using is a transfected CHO K1, GS CHO or CHO-S™ cell line
• Maximum batch culture cell densities and protein titers are needed
• You have time to adapt cells into a new medium in an effort to maximize titers
• Cell health at harvest is a priority for downstream processing

Consider CD OptiCHO™ medium when:
• You are using a transfected CHO cell line other than CHO K1, GS CHO or CHO-S™ cell lines
• Optimization of feeding strategies will be part of your base medium selection testing in an effort to maximize productivity
• Your CHO cell line is “finicky,” i.e. hard to grow or adapt

Consider CD DG44 medium when:
• You use parental DG44 or DXB11 CHO cells (dhfr-)

Liquid Media Formulations
Gibco™ AGT™ media are also available in a liquid format that offers the same performance and formulation as the AGT™ media format. Gibco™ liquid media are manufactured animal origin-free with the same complete, serum-free, protein-free, chemically-defined formulation and performance as their AGT™ media counterparts.

Design Your Own Media Specific to Your Needs
Whether it’s a bench-level experiment or large scale biomanufacturing process, we offer specialized formats, packaging, and quality levels specific to your needs. With Gibco™ cGMP Media Custom Services you can put the manufacturing power and years of technical experience of our R&D and Bioproduction Application Specialists to work for you.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Cell LineCHO
Concentration1 X
Culture EnvironmentCO2
Endotoxin LevelLow
FormatRopak Bucket
Glucose Concentration6.33 g/L
Included AntibioticsNo antibiotics
Inorganic SaltsCalcium, Magnesium, Sodium Phosphate
Product LineAGT, Gibco
Product TypeCD CHO Medium
Purity or Quality GradeCertified
Quantity100 L
Shipping ConditionDry Ice
ClassificationAnimal Origin-free, Chemically-defined, Protein-free, Serum-free
Culture TypeSuspension Cell Culture
FormPowder (AGT)
Serum LevelSerum-free
SterilityNon-sterile
With AdditivesHigh Glucose, Sodium Pyruvate, Sodium Bicarbonate
Without AdditivesNo Glutamine, No HEPES, No Phenol Red, No Thymidine, No Hypoxanthine
Unit SizeEach
Contents & Storage
Store at 2 to 8°C, in the dark.

Frequently asked questions (FAQs)

What are the main benefits of using AGT?

Complete formulations
- Homogeneous distribution of minute components
- Complete complex formulations in a dry format, including serum-free, protein-free, and chemically defined nutrient media
- Pre-adjusted to the appropriate pH and osmolality
- Require only standard supplementation with L-glutamine for L-glutamine dependent systems
- Can be customized for modified catalog and customers' proprietary formulations

Granular format
- Dissolves instantly for faster media preparation times
- Dust-free granules reduce mess and environmental contamination
- Performance equivalent to liquid media
- Workflow improvement
- Helps reduce total cycle costs due to lower number of components, decreasing time involved in raw material planning, procurement, and testing
- Faster and easier media preparation, with less potential for error
- Easy to scale up from research through production, in batch sizes from 2 kg to 6,000 kg
- Available in various packaging options

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

What is AGT?

Granulated cell culture media is an easy-to-use, granular form of dry media that is produced by applying fluid bed granulation technology to cell culture nutrient media manufacturing. The patented process allows production of complete formulations of a variety of serum-free, protein-free, and chemically defined media in a dry format.

AGT offers the benefits of liquid media without the cost, storage, and transportation issues. It is a complete medium that is pH and osmolality pre-adjusted. The granules dissolve instantly for faster media preparation time than conventional dry powder media. AGT media can therefore help reduce total cycle costs, decreasing time involved in raw material planning, procurement, and testing as well as media preparation. These benefits are all realized while maintaining comparable cell growth to liquid media.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.