Thermo Scientific™
Slide Mailers and Envelopes
 Related applications:
Anatomical Pathology
Hold slides with Thermo Scientific™ Slide Mailers and Envelopes, available in plastic or cardboard.
Error loading your content!
  Catalog number Price ({{currency}}) Availability Qty
{{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}} Add To Cart
{{addItemToList.successMessage}} {{addItemToList.listName}}
Back to top
Description
  • Plastic mailer holds five slides in a single-piece design with built-in slide separators and a hinged, tight-fitting lid
  • Cardboard mailer holds two slides
Documents
Manuals & protocols