Iron(III) chloride hexahydrate, 99+%, extra pure
Iron(III) chloride hexahydrate, 99+%, extra pure
Iron(III) chloride hexahydrate, 99+%, extra pure
Thermo Scientific Chemicals

Iron(III) chloride hexahydrate, 99+%, extra pure

CAS: 10025-77-1 | Cl3FeH12O6 | 270.29 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
2.5 kg
5 kg
25 kg
Packaging:
View all
Plastic bottle
Catalog number 125030025
Price (USD)
134.65
Online exclusive
149.00
Save 14.35
Each
Quantity:
2.5 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
134.65
Online exclusive
149.00
Save 14.35
Each
Chemical Identifiers
CAS10025-77-1
IUPAC Nameiron(3+) hexahydrate trichloride
Molecular FormulaCl3FeH12O6
InChI KeyNQXWGWZJXJUMQB-UHFFFAOYSA-K
SMILESO.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Fe+3]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow to orange to brown
Appearance (Form)Granules or lumps
Titration Iodometric99.0 to 102.0 %
Insoluble matter=<0.1 % (10 g in 50 ml H2O:HCl (49:1))
Acidity (HCl)=<0.8 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Lewis acid
RUO – Research Use Only