L(+)-Lactic acid, 20 wt.% solution in water
L(+)-Lactic acid, 20 wt.% solution in water
L(+)-Lactic acid, 20 wt.% solution in water
Thermo Scientific Chemicals

L(+)-Lactic acid, 20 wt.% solution in water

CAS: 79-33-4 | C3H6O3 | 90.078 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
Packaging:
Glass bottle
Catalog number 125070010
Price (USD)
82.65
Online exclusive
91.70
Save 9.05
Each
Quantity:
1 L
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
82.65
Online exclusive
91.70
Save 9.05
Each
Chemical Identifiers
CAS79-33-4
IUPAC Name(2S)-2-hydroxypropanoic acid
Molecular FormulaC3H6O3
InChI KeyJVTAAEKCZFNVCJ-REOHCLBHSA-N
SMILESC[C@H](O)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Appearance (Form)Solution
Titration with NaOH>=18 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only