Lithium tri-tert-butoxyaluminohydride, 93-98%
Lithium tri-tert-butoxyaluminohydride, 93-98%
Lithium tri-tert-butoxyaluminohydride, 93-98%
Thermo Scientific Chemicals

Lithium tri-tert-butoxyaluminohydride, 93-98%

CAS: 17476-04-9 | C12H28AlLiO3 | 254.27 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number 125160250
Price (TWD)
-
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS17476-04-9
IUPAC Namealuminium(3+) tris(2-methylpropan-2-olate) lithium
Molecular FormulaC12H27AlLiO3
InChI KeyHTBVGZAVHBZXMS-UHFFFAOYSA-N
SMILES[Li].[Al+3].CC(C)(C)[O-].CC(C)(C)[O-].CC(C)(C)[O-]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light grey
Appearance (Form)Powder
Infrared spectrumConforms
Assay>=93 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagent
RUO – Research Use Only