3,5-Lutidine, 98+%
3,5-Lutidine, 98+%
3,5-Lutidine, 98+%
Thermo Scientific Chemicals

3,5-Lutidine, 98+%

CAS: 591-22-0 | C7H9N | 107.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
500 mL
Packaging:
Glass bottle
Catalog number 125211000
Price (USD)
44.65
Special offer
Online exclusive
Ends: 28-Dec-2026
52.80
Save 8.15
Each
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
44.65
Special offer
Online exclusive
Ends: 28-Dec-2026
52.80
Save 8.15
Each
Chemical Identifiers
CAS591-22-0
IUPAC Name3,5-dimethylpyridine
Molecular FormulaC7H9N
InChI KeyHWWYDZCSSYKIAD-UHFFFAOYSA-N
SMILESCC1=CC(C)=CN=C1
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=98.0 %
Appearance (Form)Liquid
Appearance (Color)Clear colorless to yellow
Infrared spectrumConforms
Refractive index1.5040 to 1.5060 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only