Maleic Acid, 99%
Maleic Acid, 99%
Maleic Acid, 99%
Thermo Scientific Chemicals

Maleic Acid, 99%

CAS: 110-16-7 | C4H4O4 | 116.07 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
1 kg
5 kg
25 kg
Packaging:
View all
Plastic bottle
Catalog number 125231000
Price (USD)
41.10
Each
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
41.10
Each
Chemical Identifiers
CAS110-16-7
IUPAC Name(2E)-but-2-enedioic acid
Molecular FormulaC4H4O4
InChI KeyVZCYOOQTPOCHFL-OWOJBTEDSA-N
SMILESOC(=O)\C=C\C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder or granules and/or chunks
Titration with NaOH98.5 to 101.5 % (on dry substance)
HPLC>=98.5 %
Infrared spectrumConforms
Appearance (Color)White
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only