(R)-(-)-Mandelic acid, 99%
(R)-(-)-Mandelic acid, 99%
(R)-(-)-Mandelic acid, 99%
Acros Organics

(R)-(-)-Mandelic acid, 99%

CAS: 611-71-2 | C8H8O3 | 152.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
100 g
Packaging:
View all
Plastic bottle
Catalog number 125320250
HSN 29163990
Price (INR)
7,243.00
Each
Quantity:
25 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (INR)
7,243.00
Each
Chemical Identifiers
CAS611-71-2
IUPAC Name(2R)-2-hydroxy-2-phenylacetic acid
Molecular FormulaC8H8O3
InChI KeyIWYDHOAUDWTVEP-SSDOTTSWSA-N
SMILESO[C@@H](C(O)=O)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light yellow to light beige
Melting point130°C to 135°C
Water=<1 % (K.F.)
Specific optical rotation-152° to -158° (20°C, 589 nm) (c=2, H2O)
Appearance (Form)Crystalline powder or crystals or flakes
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only