Melatonine, 99%
Melatonine, 99%
Melatonine, 99%
Thermo Scientific Chemicals

Melatonine, 99%

Melatonine, 99%, CAS # 73-31-4, is a hormone usually produced in the pineal gland and released into the blood.
Have Questions?
Catalog NumberQuantityPackaging
1253600101 gGlass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 125360010
Price (USD)
87.10
Each
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
87.10
Each
Chemical Identifiers
CAS73-31-4
IUPAC NameN-[2-(5-methoxy-1H-indol-3-yl)ethyl]acetamide
Molecular FormulaC13H16N2O2
InChI KeyDRLFMBDRBRZALE-UHFFFAOYSA-N
SMILESCOC1=CC=C2NC=C(CCNC(C)=O)C2=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Melting point116°C to 120°C
Loss on drying=<0.5 % (1 g, 105°C)
Appearance (Form)Powder
Appearance (Color)White to cream to yellow
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

• Melatonine is a hormone produced by the pineal gland that shows several biological activities, including the somnolence, sleep-wake cycle, circadian rhythm, stress response, aging, and immunity activities

• It shows antioxidant, autocoid, paracoid, hormonal, anti- inflammatory and anti-apoptotic activities

Applications

• Melatonin shows neurohormonal activity and can upregulate antioxidant enzymes and inhibit rat NOS1

• Melatonin has been reported to be an activator of MEL-1A-R and MEL-1B-R

• The production and secretion of melatonine are dependent on beta-adrenergic receptor function

• During melatonine synthesis, the essential amino acid L-tryptophan is used as a precursor

• This compound inhibits apoptosis in immune cells and neurons, and enhances apoptotic cell death of cancer cells

• It can inhibit proliferation and metastasis of breast cancer cells by modulating Rho-associated kinase protein-1 expression

RUO – Research Use Only

General References:

  1. Borin, T.F.; Arbab, A.S.; Gelaleti, G.B.; Ferreira, L.C.; Moschetta, M.G.; Jardim-Perassi, B.V.; Iskander, A.S.; Varma, N.R.; Shankar, A.; Coimbra, V.B.; Fabri, V.A.; de Oliveira, J.G.; Zuccari, D.A. Melatonin decreases breast cancer metastasis by modulating Rho-associated kinase protein-1 expression. J Pineal Res. 2016, 60, (1), 3-15.
  2. Tan, D.X.; Manchester, L.C.; Hardeland, R.; Lopez-Burillo, S.; Mayo, J.C.; Sainz, R.M.; Reiter, R.J. Melatonin: a hormone, a tissue factor, an autocoid, a paracoid, and an antioxidant vitamin. J Pineal Res. 2003, 34, (1), 75-78.
  3. Dubocovich, M.L. Pharmacology and function of melatonin receptors. FASEB J. 1988, 2, (12), 2765-73.
  4. Reiter, R.J., et al. Free radical-mediated molecular damage. Mechanisms for the protective actions of melatonin in the central nervous system. Ann. N.Y. Acad. Sci. 2001, 939, 200-15.