LR Clonase™ II Plus enzyme
Invitrogen™
LR Clonase™ II Plus enzyme
Mix contains a proprietary blend of Int (Integrase), IHF (Integration Host Factor), and Xis (Excisionase) enzymes.
Have Questions?
보기 방식 변경buttonViewtableView
카탈로그 번호수량
1253812020 Reactions
12538200100 Reactions
카탈로그 번호 12538120
제품 가격(KRW)
1,013,400
온라인 행사
Ends: 30-Sep-2026
1,126,000
할인액 112,600 (10%)
Each
수량:
20 Reactions
제품 가격(KRW)
1,013,400
온라인 행사
Ends: 30-Sep-2026
1,126,000
할인액 112,600 (10%)
Each
Gateway™ LR Clonase™ enzyme mix contains a proprietary blend of Int (Integrase), IHF (Integration Host Factor) and Xis (Excisionase) enzymes that catalyze the in vitro recombination between an Entry clone (containing a gene of interest flanked by attL sites) and a Destination vector (containing attR sites) to generate your expression clone. We offer different formats of LR Clonase enzyme mixtures, depending on your application and desired format.
LR Clonase II Plus enzyme mix is the latest version that offers the highest recombination efficiency available(Table 1), and is specifically optimized for both single and multiple fragment cloning (Table 1). LR Clonase II Plus enzyme mix is provided in an optimized single mix of enzyme and reaction buffer, ensuring enzyme stability and easy of use with few pippetting steps for both single and multiple fragment cloning. Our LR Clonase II enzyme mix is also available in a single mix format, while the original LR Clonase® enzyme is the enzyme and buffer in separate tubes.
For Research Use Only. Not for use in diagnostic procedures.
사양
함께 사용가능한 버퍼Reaction Buffer
제품 유형LR Clonase Enzyme
수량20 Reactions
배송 조건Dry Ice
효소LR Clonase
제품라인Clonase, Gateway
Unit SizeEach
구성 및 보관
Includes LR Clonase II Plus enzyme and proteinase K solution (2 μg/μL). Store at -80°C. Guaranteed stable for 6 months when properly stored.
자주 묻는 질문(FAQ)
What is Gateway Cloning Technology?
Gateway Cloning Technology is an easy-to-use system for cloning and subcloning DNA segments (e.g. genes of interest), facilitating gene functional analysis, protein expression, and the integration of technology platforms. One can also readily clone PCR products into so-called Gateway "Entry" vectors. To shuttle inserts from one vector to another, the Gateway Cloning Technology uses bacteriophage lambda-based site-specific recombination. There is no need to use restriction enzymes and ligase to subclone inserts.
One advantage of Gateway Cloning Technology is that genes present in a single Gateway Entry vector can be subcloned into multiple different Gateway Destination vectors. After this 1 hour in vitro subcloning reaction, a high percentage of the colonies obtained carry the desired expression clone. For more details, please see the product manual for cat# 12535029 or cat# 12535037.