Specific optical rotation-2° to +2° (20°C, 589 nm) (c=10, EtOH)
Appearance (Form)Crystalline solid
Appearance (Color)Colorless to white
Infrared spectrumConforms
Melting point32°C to 36°C
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.