Mercaptoacetic Acid, 98%
Mercaptoacetic Acid, 98%
Mercaptoacetic Acid, 98%
Thermo Scientific Chemicals

Mercaptoacetic Acid, 98%

CAS: 68-11-1 | C2H4O2S | 92.11 g/mol
数量:
10 mL
1 L
2.5 L
パッケージ:
Glass bottle
製品番号(カタログ番号) 125430025
価格(JPY)
-
数量:
2.5 L
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS68-11-1
IUPAC Name2-sulfanylacetic acid
Molecular FormulaC2H4O2S
InChI KeyCWERGRDVMFNCDR-UHFFFAOYSA-N
SMILESOC(=O)CS
さらに表示
Appearance (Color)Clear colorless
Appearance (Form)Liquid
Infrared spectrumConforms
Titration Iodimetric97.5 to 102.5 %
Refractive index1.5020 to 1.5070 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only