3-Mercaptopropionic acid, 98%
3-Mercaptopropionic acid, 98%
3-Mercaptopropionic acid, 98%
Thermo Scientific Chemicals

3-Mercaptopropionic acid, 98%

CAS: 107-96-0 | C3H6O2S | 106.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
500 mL
2.5 L
Catalog number 125530025
Price (USD)
-
Quantity:
2.5 L
Request bulk or custom format
Chemical Identifiers
CAS107-96-0
IUPAC Name3-sulfanylpropanoic acid
Molecular FormulaC3H6O2S
InChI KeyDKIDEFUBRARXTE-UHFFFAOYSA-N
SMILESOC(=O)CCS
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
Appearance (Form)Liquid
Infrared spectrumConforms
Refractive index1.4910 to 1.4940 (20°C, 589 nm)
Titration with NaOH97.5 to 102.5 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagent
RUO – Research Use Only