Mesitaldehyde, 97%
Mesitaldehyde, 97%
Mesitaldehyde, 97%
Thermo Scientific Chemicals

Mesitaldehyde, 97%

CAS: 487-68-3 | C10H12O | 148.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Packaging:
Glass bottle
Catalog number 125570100
Price (USD)
91.30
Each
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
91.30
Each
Chemical Identifiers
CAS487-68-3
IUPAC Name2,4,6-trimethylbenzaldehyde
Molecular FormulaC10H12O
InChI KeyHIKRJHFHGKZKRI-UHFFFAOYSA-N
SMILESCC1=CC(C)=C(C=O)C(C)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
Infrared spectrumConforms
Refractive index1.5520 to 1.5540 (20°C, 589 nm)
GC>=96.0 %
Appearance (Form)Liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only