PB-MAX™ Karyotyping Medium
Gibco™

PB-MAX™ Karyotyping Medium

Green features
Fully supplemented medium developed for short-term culture of peripheral blood lymphocytes for cytogenetic studies and in vitro diagnostic procedures
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
12557021500 mL
12557013100 mL
Catalog number 12557021
Price (USD)
347.00
Each
Quantity:
500 mL
Price (USD)
347.00
Each

Gibco™ PB-MAX™ Karyotyping Medium is a fully-supplemented medium developed for the short-term culture of peripheral blood lymphocytes for cytogenetic studies and in vitro diagnostic procedures.

Gibco™ PB-MAX™ Karyotyping Medium features:

  • Ready-to-use format
  • Quality and performance testing
  • Classic formulation

Ready-to-use format

Gibco™ PB-MAX™ Karyotyping Medium is a frozen, 1X medium, ready to use upon thawing. No supplementation is required. Thawed medium can be stored at 2–8°C for up to 14 days.

Quality and performance testing

Every lot of Gibco™ PB-MAX™ Karyotyping Medium is performance tested by a certified US reference cytogenetics laboratory to ensure consistently superior performance. Peripheral blood lymphocytes from a normal adult donor are cultured for 72 hours in Gibco™ PB-MAX™ Karyotyping Medium before measuring the mitotic index and chromosome banding resolution. In addition, each lot is tested for sterility, pH, osmolality, and mycoplasma.

Classic formulation

Gibco™ PB-MAX™ Karyotyping Medium is an optimized RPMI 1640 medium, supplemented with Fetal Bovine Serum (FBS), L-glutamine, and phytohemagglutinin (PHA). Gibco™ PB-MAX™ Karyotyping Medium is based on formulations referenced in the Association for Cytogenetic Technologists Laboratory Manual for the culture of peripheral blood lymphocytes for cytogenetic analysis. The complete formulation is available.

Product Use

For in vitro diagnostic use.

cGMP Manufacturing and Quality System

Gibco™ PB-MAX™ Karyotyping Medium is manufactured at a cGMP compliant facility, located in Grand Island, New York. The facility is registered with the FDA as a medical device manufacturer and is certified to ISO 13485 standards.

For in vitro diagnostic use.
Specifications
Chemical Name or MaterialKaryotyping Medium
ColorRed
Green FeaturesACT Ecolabel (Silver)
IncludesL-Glutamine
Recommended StorageStorage conditions: -5 to -20°C. Protect from light. Shipping conditions: Frozen Shelf life: 12 months from date of manufacture
SterilitySterile-filtered
Concentration1X
For Use With (Application)Cell Viability; Proliferation & Function; Clinical & Translational Research; Mammalian Cell Culture
Physical FormLiquid
Product LinePB-MAX
Quantity500 mL
Unit SizeEach