2-Mesitylenesulfonyl chloride, 99%
2-Mesitylenesulfonyl chloride, 99%
2-Mesitylenesulfonyl chloride, 99%
Thermo Scientific Chemicals

2-Mesitylenesulfonyl chloride, 99%

CAS: 773-64-8 | C9H11ClO2S | 218.7 g/mol
数量:
25 g
100 g
500 g
パッケージ:
Plastic bottle
製品番号(カタログ番号) 125590250
価格(JPY)
-
数量:
25 g
パッケージ:
Plastic bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS773-64-8
IUPAC Name2,4,6-trimethylbenzene-1-sulfonyl chloride
Molecular FormulaC9H11ClO2S
InChI KeyPVJZBZSCGJAWNG-UHFFFAOYSA-N
SMILESCC1=CC(C)=C(C(C)=C1)S(Cl)(=O)=O
さらに表示
Appearance (Color)White to grey
Infrared spectrumConforms
Titration Argentometric>=98.5 % (ex Cl)
Appearance (Form)Crystalline powder
Melting point53°C to 57°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only