Methanesulfonic acid, 99%, extra pure
Methanesulfonic acid, 99%, extra pure
Methanesulfonic acid, 99%, extra pure
Thermo Scientific Chemicals

Methanesulfonic acid, 99%, extra pure

CAS: 75-75-2 | CH4O3S | 96.1 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 mL
250 mL
1 L
5 L
Packaging:
View all
Glass bottle
Catalog number 125610010
Price (USD)
221.65
Online exclusive
246.00
Save 24.35
Each
Quantity:
1 L
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
221.65
Online exclusive
246.00
Save 24.35
Each
Chemical Identifiers
CAS75-75-2
IUPAC Namemethanesulfonic acid
Molecular FormulaCH4O3S
InChI KeyAFVFQIVMOAPDHO-UHFFFAOYSA-N
SMILESCS(O)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Color scale=<150 APHA
Titration with NaOH>=98.5 %
Infrared spectrumConforms
Appearance (Form)Clear liquid
Refractive index1.4285 to 1.4315 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only