2'-Methoxyacetophenone, 99%
2'-Methoxyacetophenone, 99%
2'-Methoxyacetophenone, 99%
Thermo Scientific Chemicals

2'-Methoxyacetophenone, 99%

CAS: 579-74-8 | C9H10O2 | 150.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
100 mL
Packaging:
Glass bottle
Catalog number 125671000
Price (USD)
204.00
Each
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
204.00
Each
Chemical Identifiers
CAS579-74-8
IUPAC Name1-(2-methoxyphenyl)ethan-1-one
Molecular FormulaC9H10O2
InChI KeyDWPLEOPKBWNPQV-UHFFFAOYSA-N
SMILESCOC1=CC=CC=C1C(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear light yellow to orange to light brown
Infrared spectrumConforms
GC>=98.5 %
Appearance (Form)Liquid
Refractive index1.5383 to 1.5403 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only