2-Methoxybenzyl alcohol, 99%
2-Methoxybenzyl alcohol, 99%
2-Methoxybenzyl alcohol, 99%
Thermo Scientific Chemicals

2-Methoxybenzyl alcohol, 99%

CAS: 612-16-8 | C8H10O2 | 138.17 g/mol
数量:
10 mL
50 mL
250 mL
パッケージ:
Glass bottle
製品番号(カタログ番号) 125760500
価格(JPY)
-
数量:
50 mL
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS612-16-8
IUPAC Name(2-methoxyphenyl)methanol
Molecular FormulaC8H10O2
InChI KeyWYLYBQSHRJMURN-UHFFFAOYSA-N
SMILESCOC1=CC=CC=C1CO
さらに表示
Appearance (Color)Clear colorless to Yellow
Appearance (Form)Liquid
Infrared spectrumConforms
Refractive index1.5460 to 1.5490 (20°C, 589 nm)
GC>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only