3-Methoxybenzyl alcohol, 98%
3-Methoxybenzyl alcohol, 98%
3-Methoxybenzyl alcohol, 98%
Thermo Scientific Chemicals

3-Methoxybenzyl alcohol, 98%

CAS: 6971-51-3 | C8H10O2 | 138.17 g/mol
Have Questions?
Catalog NumberQuantityPackaging
12577025025 mLGlass bottle
Catalog number 125770250
Price (EUR)
62,30
Each
Quantity:
25 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
62,30
Each
Chemical Identifiers
CAS6971-51-3
IUPAC Name(3-methoxyphenyl)methanol
Molecular FormulaC8H10O2
InChI KeyIIGNZLVHOZEOPV-UHFFFAOYSA-N
SMILESCOC1=CC=CC(CO)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow to light brown
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only