GeneBLAzer® In Vitro Detection Kit
Catalog number: 12578126
Invitrogen™  Related applications: Cellular Imaging | Protein Assays and Analysis
Error loading your content!
  Catalog number
Select a plan
Unit size
Price ({{currency}}) Your price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{subscriptionDetails.selectedRatePlan.billingPeriod}} {{subscriptionDetails.selectedRatePlan.billingPeriod}} {{product.availability.message}}
Pro add-ons
Your on-site stock
›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top
The GeneBLAzer™ In Vitro Detection Kit is designed to measure β-lactamase reporter activity in mammalian cell lysates, using the β-lactamase substrate CCF2-FA.
For Research Use Only. Not for use in diagnostic procedures.
BrazilCouponProductLists: Dealers_40
Substrate: CCF2-FA
Vector Type: GeneBLAzer Vectors
Product Size: 115 reactions
Substrate Type: Beta-Lactamase Substrate
Detection Method: Fluorescent
Substrate Properties: Chemical Substrate
Contents & storage
In vitro detection kits contain 100 µg of CCF2-FA substrate (enough for 115 96-well reactions). Substrate should be stored at -20°C, dessicated, and protected from light. All reagents are guaranteed stable 6 months when properly stored.
Manuals & protocols
Recommended products