Product Image
Invitrogen™

Optifect™ Transfection Reagent

Optifect™ Transfection Reagent is a pre-optimized formulation designed to ensure highly efficient gene delivery to a wide variety of cell자세히 알아보기
Have Questions?
카탈로그 번호수량
125790171 mL
카탈로그 번호 12579017
제품 가격(KRW)
843,000
온라인 행사
Ends: 31-Dec-2026
991,000
할인액 148,000
Each
수량:
1 mL
제품 가격(KRW)
843,000
온라인 행사
Ends: 31-Dec-2026
991,000
할인액 148,000
Each
Optifect™ Transfection Reagent is a pre-optimized formulation designed to ensure highly efficient gene delivery to a wide variety of cell lines transfected at low confluence or density (10%-70%) (Figure 1). Optifect™ Reagent can also be used with cells that are sensitive to Lipofectamine™ 2000 or other commercially available transfection reagents. Optifect™ Reagent facilitates high levels of protein expression from sparsely plated cells (Table 1). Useful in applications such as studies of cell surface proteins, post-transfection proliferation assays, or other situations where cells must be allowed to grow significantly following transfection. Optifect™ has been shown to be less toxic to cells compared to other commercially available transfection reagents.
For Research Use Only. Not for use in diagnostic procedures.
사양
용도(애플리케이션)Transfection
고처리량 호환성High-throughput Compatible
제품라인Optifect
제품 유형Transfection Reagent
수량1 mL
혈청 호환 가능Yes
배송 조건Wet Ice
셀 유형Established Cell Lines, Primary Cells, Hard-to-Transfect Cells
형식6-well Plate, 12-well Plate, 24-well Plate, 48-well Plate, 96-well Plate, Flasks
샘플 종류Plasmid DNA
Transfection TechniqueLipid-based Transfection
VectorpLATE
Unit SizeEach
구성 및 보관
One unit vial contains of reagent. Store at +4°C. Guaranteed stable for 6 months when properly stored.

자주 묻는 질문(FAQ)

I accidentally left my lipid reagent at room temperature. Can I still use it?

Yes, all of our lipid transfection reagents are stable at room temperature for months.

Find additional tips, troubleshooting help, and resources within our Lipid-Based Transfection Support Center.

What is the difference between reverse transfection and forward transfection? What should I use?

In forward transfection, cells are seeded to appropriate confluence or cell density in wells or dishes, and the lipid-DNA complexes are added the next day. In reverse transfection, the transfection complexes are prepared inside the wells, after which cells and medium are added. Reverse transfection is faster to perform than forward transfection, and is the method of choice for high-throughput transfection. For non-high-throughput transfections, generally forward transfections have better efficiency for most cell types.

Find additional tips, troubleshooting help, and resources within our Transfection Support Center.

Is there a place where I can find references from other researchers who have used your transfection reagents?

Visit the product page for each reagent type and you will see a list of references at the bottom of the page. A table that lists specific cell line references is also accessible. We also recommend www.highwire.org as a search engine to find a large selection of up-to-date research articles using our transfection products. Simply include the name of the transfection reagent and your cell line/application of interest in your search criteria.

Find additional tips, troubleshooting help, and resources within our Transfection Support Center.

Can I use antibiotics in the medium during transfection?

Antibiotics can be used in the medium for culturing of cell lines. However, we do not recommend using antibiotics in the transfection medium unless previously tested in the cell type and payload being transfected. This is because presence of antibiotics during transfection may adversely affect transfection efficiency (i.e., positively charged antibiotics binding to the DNA being transfected) and overall health of cells being transfected.

For stable transfection, we recommend waiting wait 24-48 hrs after transfection before adding selected antibiotics.

Find additional tips, troubleshooting help, and resources within ourTransfection Basics Support Center.

Is it necessary to use serum-free medium during lipid transfection?

It is not necessary to use serum-free medium during lipid transfection. However, it is critical to form the lipid:nucleic acid complex in the absence of serum, because proteins can interfere with complex formation. Once the complexes are formed, they can be added to cells in serum-containing medium. For optimal results with Lipofectin Transfection Reagent, we recommend performing transfection in medium without serum.

Find additional tips, troubleshooting help, and resources within our Lipid-Based Transfection Support Center.