3-Methoxy-1-butanol, 99%
3-Methoxy-1-butanol, 99%
3-Methoxy-1-butanol, 99%
Thermo Scientific Chemicals

3-Methoxy-1-butanol, 99%

CAS: 2517-43-3 | C5H12O2 | 104.15 g/mol
数量:
100 mL
1 L
パッケージ:
Glass bottle
製品番号(カタログ番号) 125791000
価格(JPY)
-
数量:
100 mL
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS2517-43-3
IUPAC Name3-methoxybutan-1-ol
Molecular FormulaC5H12O2
InChI KeyJSGVZVOGOQILFM-UHFFFAOYNA-N
SMILESCOC(C)CCO
さらに表示
Infrared spectrumConforms
Appearance (Color)Clear colorless
Water=<0.2 %
Refractive index1.4150 to 1.4165 (20°C, 589 nm)
Appearance (Form)Liquid
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only