Platinum® Blue PCR SuperMix 96, non-skirted - FAQs
Platinum® Blue PCR SuperMix 96, non-skirted - FAQs
View additional product information for Platinum® Blue PCR SuperMix 96, non-skirted - FAQs (12580049)
1 product FAQs found
What is the difference between Platinum technology and AccuPrime technology?
With Platinum technology, anti-DNA polymerase antibodies bind to the enzyme until the denaturing step at 94 degrees C, when the antibodies degrade. The polymerase is now active and primer extension can occur. AccuPrime Taq combines Platinum Taq (Taq + Platinum antibodies) with proprietary thermostable AccuPrime accessory proteins. The 10X reaction buffer contains the accessory proteins which enhance specific primer-template hybridization during each cycle of PCR.