B-27™ Supplement (50X), minus vitamin A, 100 mL - FAQs
B-27™ Supplement (50X), minus vitamin A, 100 mL - FAQs
View additional product information for B-27™ Supplement (50X), minus vitamin A - FAQs (12587010, 12587001)
6 product FAQs found
Can B-27 supplement be filtered?
Yes, the protein content in this product is high enough so filtering through a low protein binding filter as a 50X or 1X in solution should not be a problem.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
Can Neurobasal Medium alone or Neurobasal Medium supplemented with B-27 supplement be frozen to extend product shelf life?
No, it is not recommended to freeze media due to the potential of precipitates forming upon thaw. Inorganic salts and amino acids in the formulation may come out of solution when exposed to temperature fluctuations. These precipitates will not go back into solution easily once formed.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
Do you offer B-27 supplements with and without insulin?
Yes, we do offer B-27 Supplement, minus insulin (Cat. Nos. A1895601 and A1895602). All our other B-27 supplements do contain insulin.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
Can I thaw B-27 Supplements in a 37 degrees C water bath or incubator?
We do not recommend thawing at 37 degrees C. Best results are observed upon thawing at room temperature for approximately 2 hrs or overnight at 2-8 degrees C.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
How do I thaw B-27 Supplements?
We recommend thawing it at 4 degrees C overnight or at room temperature for 2 hours. We don't recommend thawing in a water bath or incubator.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
Can I re-freeze B-27 Supplements into smaller aliquots for future use?
Yes, we recommend that you aliquot when you first thaw it and thereafter subject the aliquot to no more than 2 additional freeze/thaw cycles.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.