5-Methoxyindole, 98%
5-Methoxyindole, 98%
5-Methoxyindole, 98%
Thermo Scientific Chemicals

5-Methoxyindole, 98%

CAS: 1006-94-6 | C9H9NO | 147.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 125870250
Price (USD)
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS1006-94-6
IUPAC Name5-methoxy-1H-indole
Molecular FormulaC9H9NO
InChI KeyDWAQDRSOVMLGRQ-UHFFFAOYSA-N
SMILESCOC1=CC=C2NC=CC2=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to brown
Appearance (Form)Powder or crystals
Infrared spectrumConforms
Melting point51°C to 57°C
GC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only