5-Methoxy-2-methylindole, 99+%
5-Methoxy-2-methylindole, 99+%
5-Methoxy-2-methylindole, 99+%
Thermo Scientific Chemicals

5-Methoxy-2-methylindole, 99+%

CAS: 1076-74-0 | C10H11NO | 161.20 g/mol
Quantity:
5 g
25 g
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 125940050
Price (JPY)
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS1076-74-0
IUPAC Name5-methoxy-2-methyl-1H-indole
Molecular FormulaC10H11NO
InChI KeyVSWGLJOQFUMFOQ-UHFFFAOYSA-N
SMILESCOC1=CC=C2NC(C)=CC2=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light beige
Appearance (Form)Crystalline powder or crystals
Infrared spectrumConforms
Melting point85°C to 88°C
GC>=99.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only