Interested in a bulk or custom version of this product? Request pricing.

CIB2 Recombinant Human Protein, N-His Tag

Catalog number:  12596H07E50

 Related applications: Protein Assays and Analysis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Calcium and Integrin Binding family member 2 (CIB2), recombinant human protein is supplied as a frozen protein. In general, recombinant proteins can be used as protein stucture analysis and in cell biology research applications.

This recombinant protein was expressed from a DNA sequence encoding the mature form of human CIB2 (O75838) (Met 1-Ile 187) fused to a polyhistide tag at the N-terminus.

Activity: This recombinant protein has not been tested.

Formulation: 50 mM Tris, 20% glycerol, pH8.0.

Reconstitution: No reconstitution needed. Store at -80°C. Avoid repeated freeze-thaw cycles.

Expiration Date: Expires one year from date of receipt when stored as instructed.



This protein is manufactured by SINO Biological.

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Expression: E. coli
Form: Liquid
Gene Aliases: KIP2
Gene ID: 10518
Gene Symbol: CIB2
Molecular Weight: 23.1 kDa
Product Line: Sino Biological®
Product Size: 50 µg
Protein Family: Other Proteins
Protein Form: Full Length, Recombinant
Protein Name: Calcium and Integrin Binding family member 2
Protein Tag or Fusion: His Tag
Purification: PAGE
Purity: ≥ 90% by SDS-PAGE
Research Category: Signal Transduction
Species: Human
Tag Position: N-Terminal
Validated Application: Assay Standard

Contents & storage

Includes 1 vial of protein in suspension buffer. Store at -80 °C.

Documents

Manuals & protocols