SuperScript IV UniPrime One-Step RT-PCR System
Invitrogen™

SuperScript IV UniPrime One-Step RT-PCR System

Invitrogen SuperScript IV UniPrime One-Step RT-PCR System is a convenient one-step formulation for performing fast cDNA synthesis and PCR amplification in a single tube.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescriptionNo. of Reactions
12597100Colored100 x 50 μL Reactions
12596025Dye-free25 x 50 μL Reactions
12597025Colored25 x 50 μL Reactions
12596100Dye-free100 x 50 μL Reactions
12596500Dye-free500 x 50 μL Reactions
12597500Colored500 x 50 μL Reactions
Catalog number 12597100
Price (MXN)
-
Description:
Colored
No. of Reactions:
100 x 50 μL Reactions
Invitrogen SuperScript IV UniPrime One-Step RT-PCR System is a convenient one-step formulation for performing fast cDNA synthesis and PCR amplification in a single tube. It combines the high-processivity SuperScript IV Reverse Transcriptase (RT) with a novel UniPrime RT-PCR Master Mix to provide easier use and unmatched performance when compared to other one-step RT-PCR kits.

The Invitrogen SuperScript IV UniPrime One-Step RT-PCR System offers a convenient one-step process  for performing fast cDNA synthesis and PCR amplification in a single tube. It combines high-processivity SuperScript IV Reverse Transcriptase (RT) with a novel UniPrime RT-PCR master mix for easier use and unmatched  performance compared to other one-step RT-PCR kits.

Highlights
• Simplified workflow—universal primer annealing at 60°C decreases optimization time
• Easy pipetting—colored buffers for visual tracking of reaction setup and direct sample loading on gels
• Suitable for automation—two-phase hot-start activation mechanism for room temperature setup and benchtop stability for up to 24 hours
• Superior performance—high yields and reliable target detection even with challenging RNA samples

Combination of superior enzymes
The SuperScript IV UniPrime One-Step RT-PCR System utilizes a combination of superior enzymes. The SuperScript IV Reverse Transcriptase mix helps produce an efficient reverse transcription step through its high thermostability, processivity, and ability to synthesize cDNA from variety of RNA samples, while its high sensitivity helps ensure reliable target detection even when RNA input is limited (down to 0.01 pg RNA).

The 2X UniPrime RT-PCR master mix contains modified Platinum SuperFi II DNA Polymerase, which delivers high specificity and high yields and is ideally suited for applications that require sequence accuracy. This novel master mix formulation enables universal primer annealing at 60°C for most primer pairs designed following the general design rules and helps minimize optimization steps, save time, and avoid mistakes in RT-PCR runs.

Two-phase hot start
The innovative two-phase hot-start mechanism helps deliver high specificity and yield in one-step RT-PCR through temporal separation of the activities of the reverse transcriptase and DNA polymerase. This not only allows reaction setup at room temperature, but also helps ensure the stability of assembled reactions for up to 24 hours for targets up to 3 kb and for up to 4h for targets >3 kb.

Colored format
The SuperScript IV UniPrime One-Step RT-PCR System is availabe in a standard dye-free format or colored format containing tracking dyes. When the red tracking dye in the SuperScript IV RT mix and the blue tracking dye in the UniPrime RT-PCR master mix are combined during reaction setup, the final reaction mix turns to purple. This allows for simple visual tracking of reaction setup and helps prevent pipetting errors. The final reaction product can also be directly loaded on agarose gels for a more streamlined workflow.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
DescriptionColored
Format1-step RT-PCR Kit
Hot StartBuilt-in Hot Start
Multiplex CapabilityYes
No. of Reactions100 x 50 μL Reactions
Optimal Reaction Temperature50°C to 65°C
Quantity100 reactions
Reaction FormatMaster Mix
Reagent TypeReverse Transcription
Reverse TranscriptaseSuperScript IV
Size (Final Product)Up to 13 kb
Starting MaterialRNA
Technique1-step RT-PCR
Detection MethodGel Electrophoresis
GC-Rich PCR PerformanceHigh
PCR Method1-step RT-PCR
Unit SizeEach
Contents & Storage
• SuperScript IV RT Mix, red (200 μL)
• 2X UniPrime RT-PCR Master Mix, blue (2 x 1.25 mL)
• Nuclease-free water (2 x 1.25 mL)

Store at -25°C to -15°C.


Frequently asked questions (FAQs)

Can I set up my reactions at room temperature using SuperScript IV UniPrime One-Step RT-PCR System?

Yes. Reaction setup with SuperScript IV UniPrime One-Step RT-PCR System can be performed at room temperature. Moreover, due to innovative two-phase hot start mechanism, preassembled reactions remain stable at room temperature for up to 24 hours for targets up to 3 kb and up to 4 hours for targets >3 kb.

What DNA Polymerase is included in SuperScript IV UniPrime One-Step RT-PCR System?

SuperScript IV UniPrime One-Step RT-PCR System includes 2X UniPrime RT-PCR Master Mix containing modified Invitrogen Platinum SuperFi II DNA Polymerase, which offers high specificity, high yields, and is ideally suited for applications that require sequence accuracy. Contrary to common proofreading enzymes, this polymerase is compatible with dUTP.

Will the tracking dyes in SuperScript IV One-Step RT-PCR System Colored interfere with fragment analysis in E-Gel agarose gels?

Red and blue tracking dyes included in the colored format of SuperScript IV One-Step RT-PCR System do

Which DNA polymerase is included in SuperScript IV UniPrime One-Step RT-PCR System?

SuperScript IV UniPrime One-Step RT-PCR System includes 2X UniPrime RT-PCR master mix containing modified Platinum SuperFi II DNA Polymerase, which provides high specificity and high yields, and is ideally suited for applications that require sequence accuracy. On the contrary to common proofreading enzymes, this polymerase is compatible with dUTP.

Find additional tips, troubleshooting help, and resources within our PCR and cDNA Synthesis Support Center.

Do the tracking dyes in the colored format of SuperScript IV UniPrime One-Step RT-PCR System interfere with electrophoresis in E-Gel agarose gels?

Red and blue tracking dyes included in the colored format of SuperScript IV UniPrime One-Step RT-PCR System do not interfere with electrophoresis in E-Gel agarose gels. Please note that the reaction product obtained with both the colored and dye-free format should be diluted 2- to 30-fold for optimal separation using E-Gel agarose gels.

Find additional tips, troubleshooting help, and resources within our PCR and cDNA Synthesis Support Center.