Product Image
Thermo Scientific™
GFCI Ground Fault Circuit Interrupter Accessory
Ensure extra electrical protection when a GFCI wall outlet is not available by using Thermo Scientific™ GFCI Ground Fault Circuit Interrupter accessory.
Have Questions?
Cambiar vistabuttonViewtableView
Número de catálogoPara utilizar con (equipo)
126000000050RTE/EX-7 y 10
126000000051RTE/EX-7, RTE-740, EX-35
Número de catálogo 126000000050
Precio (CLP)
-
Para utilizar con (equipo):
RTE/EX-7 y 10
Ensure extra electrical protection when a GFCI wall outlet is not available by using Thermo Scientific™ GFCI Ground Fault Circuit Interrupter accessory.
  • For use with RTE or EX series bath chillers
Ordering Alerts:
Can only be used with 115V 60Hz units
  • Warranty varies by country, please contact us for details
  • Especificaciones
    Para utilizar con (equipo)RTE/EX-7 y 10
    Requisitos eléctricos115 V 60 Hz
    TipoGFCI
    Unit SizeEach