4-Methoxyphenol, 99%
4-Methoxyphenol, 99%
4-Methoxyphenol, 99%
Thermo Scientific Chemicals

4-Methoxyphenol, 99%

CAS: 150-76-5 | C7H8O2 | 124.14 g/mol
数量:
100 g
500 g
1 kg
5 kg
25 kg
製品番号(カタログ番号) 126000051
価格(JPY)
-
数量:
5 kg
一括またはカスタム形式をリクエストする
化学物質識別子
CAS150-76-5
Appearance (Color)White to light yellow
Appearance (Form)Flakes
Infrared spectrumConforms
Melting point54°C to 57°C
GC>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only