4-Methoxyphenylacetic acid, 99%
4-Methoxyphenylacetic acid, 99%
4-Methoxyphenylacetic acid, 99%
Thermo Scientific Chemicals

4-Methoxyphenylacetic acid, 99%

CAS: 104-01-8 | C9H10O3 | 166.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
250 g
1 kg
Packaging:
Plastic bottle
Catalog number 126022500
Price (USD)
-
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS104-01-8
IUPAC Name2-(4-methoxyphenyl)acetic acid
Molecular FormulaC9H10O3
InChI KeyNRPFNQUDKRYCNX-UHFFFAOYSA-N
SMILESCOC1=CC=C(CC(O)=O)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Titration with NaOH>=98.5 %
Appearance (Color)White to light yellow
Appearance (Form)Crystalline powder and/or chunks
Infrared spectrumConforms
Melting point84°C to 88°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only