Try new Gibco Insulin-Transferrin-Selenium Select supplements, an animal-origin free formulation. Available in ITS-G & ITS-X. Learn more >
Try TrypLE Express Enzyme (1X), no phenol red for free with the Aspire member program
TrypLE™ Express Enzyme (1X), no phenol red
Gibco™

TrypLE™ Express Enzyme (1X), no phenol red

Green features
Unlike porcine trypsin, free of animal-derived components
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
12604013100 mL
12604021500 mL
1260403920 x 100mL
126040545 L
Catalog number 12604013
Price (USD)
40.65
Online exclusive
44.50
Save 3.85
Each
Quantity:
100 mL
Customize this product
Price (USD)
40.65
Online exclusive
44.50
Save 3.85
Each

TrypLE Express is an animal origin-free, recombinant enzyme used for dissociating a wide range of adherent mammalian cells, including CHO, HEK 293, A529, primary human keratinocytes, and embryonic stem cells. TrypLE Express cleaves peptide bonds on the C-terminal sides of lysine and arginine, and is a direct replacement for trypsin. Its exceptional purity increases specificity and reduces damage to cells that can be caused by other enzymes present in some trypsin extracts. We offer a variety of TrypLE formulations for a range of cell culture applications.

Compared to trypsin and other dissociation reagents, TrypLE Express is:

  • Gentle on cells
  • Room temperature-stable
  • Easy to use
  • Animal origin-free (AOF)

This TrypLE Express is modified as follows:

With: EDTA

Without: Phenol Red

The formulation of TrypLE solutions is available, although we keep the enzyme concentration confidential.

Gentle on cells

TrypLE Express' exceptional purity increases specificity due to the action of a single enzyme. This reduces damage caused by cleavage from multiple enzymes in trypsin and other extracts.

Room temperature-stable

TrypLE Express remains stable for 24 months at room temperature or 2–8°C, making storage and handling easy and convenient.

Easy to use

TrypLE Express can be directly substituted for trypsin in existing protocols. In addition, dilution alone inactivates TrypLE Express, avoiding the need for trypsin inhibitors, such as FBS.

Animal origin-free (AOF)

Unlike porcine trypsin, TrypLE Express is free of animal-derived components.

cGMP manufacturing and quality system

For supply chain continuity, we manufacture Gibco TrypLE Express Enzyme (1X), no phenol red at two separate facilities located in Grand Island, NY and Scotland, UK. Both sites are compliant with cGMP manufacturing requirements and are certified to the ISO 13485 standard.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Cell TypeMammalian Cells
ChelatorsEDTA
Concentration1 X
Culture TypeAdherent Cell Culture
Green FeaturesACT Ecolabel (Silver)
OsmolalityCheck and Record
Product LineGibco
Quantity100 mL
RemarksRoom Temperature Stable
Shelf Life24 Months
Shipping ConditionRoom Temperature
SourceAnimal origin-free
Tested ForIn vitro Bioassay
ClassificationAnimal Origin-free
FormLiquid
Product TypeCell Culture Dissociation Reagent
SterilitySterile-filtered
Without AdditivesNo Phenol Red
pH6 to 8
Unit SizeEach
Contents & Storage
Storage conditions: 15°C to 30°C, protect from light
Alternate storage conditions: 2°C to 8°C or -5°C to -20°C
Shipping conditions: Room temperature
Shelf life: 24 months from date of manufacture

Frequently asked questions (FAQs)

How can I assess residual TrypLE reagent remaining after my downstream processing steps?

We offer the Residual TrypLE ELISA Kit (Cat. No. A6533101) that can specifically detect the recombinant protease (rProtease) in TrypLE reagent.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

Does Diploid Growth Serum-Reduced Medium (SRM) contain L-glutamine?

The basal Diploid Growth Serum-Reduced Medium already contains 6 mM L-glutamine.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

Do I have to adjust my infection parameters when using Diploid Production Serum-Free Medium (SFM)?

We recommend evaluating performance with Diploid Production Serum-Free Medium using current conditions, however, multiplicity of infection, time of infection, and time of harvest may be different than with conventional media. Closely monitor cells for cytopathic effect and evaluate viral titers at multiple time points.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

Can Diploid Growth Serum-Reduced Medium (SRM) be supplemented with different types of serum?

Diploid Growth Serum-Reduced Medium can be supplemented with Fetal Bovine Serum, Newborn Calf Serum, Donor Bovine Serum with Iron, or Bovine Serum.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

Which cell lines can I culture in Diploid Growth Serum-Reduced Medium (SRM)?

Diploid Growth Serum-Reduced Medium has been evaluated with fibroblast cell lines, including MRC-5, WI-38, IMR-90, BS-2, and CEF cells.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

Citations & References (3)

Citations & References
Abstract
Method for Large-scale Production of hIPSC Spheroids.
Authors:Lemarié L,Courtial EJ,Sohier J
Journal:Bio-protocol
PubMed ID:38618177
Stem cell spheroids are rapidly becoming essential tools for a diverse array of applications ranging from tissue engineering to 3D cell models and fundamental biology. Given the increasing prominence of biotechnology, there is a pressing need to develop more accessible, efficient, and reproducible methods for producing these models. Various techniques ... More
A simple tool to improve pluripotent stem cell differentiation.
Authors:Chetty S, Pagliuca FW, Honore C, Kweudjeu A, Rezania A, Melton DA,
Journal:Nat Methods
PubMed ID:23584186
We describe a method to help overcome restrictions on the differentiation propensities of human pluripotent stem cells. Culturing pluripotent stem cells in dimethylsulfoxide (DMSO) activates the retinoblastoma protein, increases the proportion of cells in the early G1 phase of the cell cycle and, in more than 25 embryonic and induced ... More
A rapid, cost-effective method for counting human embryonic stem cell numbers as clumps.
Authors:Prowse A, Wolvetang E, Gray P,
Journal:Biotechniques
PubMed ID:19594444
Enumeration of human embryonic stem cell (hESC) numbers through single cell digestion can be time consuming especially in high-throughput or multi-factorial analysis containing 50+ samples. We have developed a reproducible, cost-effective method of counting hESCs in clumps circumventing the need to manually dissociate each sample to single cells. The method ... More