TrypLE™ Express Enzyme (1X), phenol red
Gibco™

TrypLE™ Express Enzyme (1X), phenol red

Green features
Animal origin-free, recombinant enzyme
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
1260503620 x 100mL
12605010100 mL
126050935 L
12605028500 mL
Catalog number 12605036
Price (MXN)
-
Quantity:
20 x 100mL
Customize this product

TrypLE Express is an animal origin-free, recombinant enzyme used for dissociating a wide range of adherent mammalian cells, including CHO, HEK 293, A549, primary human keratinocytes, and embryonic stem cells. TrypLE Express cleaves peptide bonds on the C-terminal sides of lysine and arginine, and is a direct replacement for trypsin. Its exceptional purity increases specificity and reduces damage to cells that can be caused by other enzymes present in some trypsin extracts.

We offer a variety of TrypLE formulations for a range of cell culture applications.

Compared to trypsin and other dissociation reagents, Gibco TrypLE Express is:

Gentle on cells

  • Room-temperature stable
  • Easy to use
  • Animal origin-free (AOF)

This TrypLE Express is modified as follows:

With: Phenol Red, EDTA

The formulation of TrypLE solutions is available, although we keep the enzyme concentration confidential.

Gentle On Cells

TrypLE Express' exceptional purity increases specificity due to the action of a single enzyme. This reduces damage caused by cleavage from enzymes present in some trypsin preparations and in other protease extracts.

Room-Temperature Stable

TrypLE Express is room-temperature stable and ready to use. TrypLE Express remains stable for 24 months at room temperature or 2–8°C, making storage and handling easy and convenient.

Easy to Use

TrypLE Express can be directly substituted for trypsin in existing protocols. In addition, dilution alone inactivates TrypLE Express, avoiding the need for trypsin inhibitors, such as FBS.

Animal Origin-Free (AOF)

Unlike porcine trypsin, TrypLE Express is free of animal-derived components.

cGMP Manufacturing and Quality System

For supply chain continuity, we manufacture Gibco TrypLE Express at two separate facilities located in Grand Island, NY and Scotland, UK. Both sites are compliant with cGMP manufacturing requirements, are certified to the ISO 13485 standard, and are registered with the FDA as medical device manufacturers.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Cell TypeMammalian Cells
ChelatorsEDTA
Concentration1 X
Culture TypeAdherent Cell Culture
Green FeaturesACT Ecolabel (Silver)
OsmolalityCheck and Record
Product LineGibco
Quantity20 x 100mL
RemarksRoom Temperature Stable
Shelf Life24 Months
Shipping ConditionRoom Temperature
SourceAnimal origin-free
Tested ForIn vitro Bioassay
ClassificationAnimal Origin-free
FormLiquid
Product TypeCell Culture Dissociation Reagent
SterilitySterile-filtered
With AdditivesPhenol Red, EDTA
Unit SizeEach
Contents & Storage
Storage conditions: 15°C to 30°C, protect from light
Alternate storage conditions: 2°C to 8°C or -5°C to -20°C
Shipping conditions: Room temperature
Shelf life: 24 months from date of manufacture

Frequently asked questions (FAQs)

What enzymes do you recommend for passaging mesenchymal stem cells (MSC)?

We recommend using TrypLE Express Enzyme or TrypLE Select Enzyme for passaging mesenchymal stem cells (MSCs). The usage of trypsin will result in lower cell viability upon passage. TrypLE Express and TrypLE Select are defined, animal origin-free, and stable at both room and refrigerated temperatures.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

Does TrypLE Express Enzyme have a His-tag?

TrypLE Express Enzyme does not have a His-tag

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

Is there an upper limit for the amount of time cells can be incubated with TrypLE Express Enzyme, TrypLE Select Enzyme, or CTS TrypLE Select Enzyme for dissociation?

There is no upper limit for TrypLE enzyme incubation. You can incubate your cells with TrypLE longer to detach cells. 30-45 min at 37 degrees C will be usually fine. Or you can consider using TrypLE Select Enzyme (10X) (Cat. No. A1217701 or A1217702 or A1217703) or CTS TrypLE Select Enzyme (Cat. No. A4738001) depending upon the application.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

For your TrypLE products, how does the enzyme get inactivated?

Our TrypLE enzymes are very gentle, and there is no need to use serum or individual proteins to inactivate the enzyme. The TrypLE enzyme activity is inactivated by dilution using the following procedure:

After cell dissociation, transfer the cell suspension to a 15-mL conical tube. Centrifuge at 100 x g for 5-10 min to remove residual enzyme activity. Discard the supernatant and resuspend the cell pellet in 2-5 mL of pre-warmed complete medium.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

What is the difference between TrypLE Express Enzyme, with and without phenol red? What does the phenol red do?

The difference between the two products is just the presence or absence of phenol red. Phenol red is a pH indicator. You can use either product to dissociate your cells. When you use TrypLE Express enzyme to dissociate cells, they very likely become single cells. If you want to passage cells as clumps, you may consider using other reagents such as EDTA.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

Citations & References (1)

Citations & References
Abstract
A simple tool to improve pluripotent stem cell differentiation.
Authors:Chetty S, Pagliuca FW, Honore C, Kweudjeu A, Rezania A, Melton DA,
Journal:Nat Methods
PubMed ID:23584186
We describe a method to help overcome restrictions on the differentiation propensities of human pluripotent stem cells. Culturing pluripotent stem cells in dimethylsulfoxide (DMSO) activates the retinoblastoma protein, increases the proportion of cells in the early G1 phase of the cell cycle and, in more than 25 embryonic and induced ... More