CD DG44 Medium
Gibco™

CD DG44 Medium

CD DG44 Medium is a chemically defined, protein-free medium optimized for the growth of dihydrofolate reductase deficient (DHFR-) Chinese HamsterRead more
Have Questions?
Catalog NumberQuantity
126100101 L
Catalog number 12610010
Price (USD)
183.00
Each
Quantity:
1 L
Price (USD)
183.00
Each
CD DG44 Medium is a chemically defined, protein-free medium optimized for the growth of dihydrofolate reductase deficient (DHFR-) Chinese Hamster Ovary (CHO) cells and expression of recombinant proteins in suspension culture. CD DG44 fully chemically defined formulation contains no lysates, hydrolysates, nor proteins or peptide components of animal, plant, or synthetic origin.

When to Use CD DG44 Medium
• You use parental DG44 or DXB11 CHO cells (dhfr-)

When to Consider Another Gibco™ Medium

Consider CD FortiCHO™ medium when:
• The cell line you are using is a transfected CHO K1, GS CHO or CHO-S™ cell line
• Maximum batch culture cell densities and protein titers are needed
• You have time to adapt cells into a new medium in an effort to maximize titers
• Cell health at harvest is a priority for downstream processing

Consider CD OptiCHO™ medium when:
• You are using a transfected CHO cell line other than CHO K1, GS CHO or CHO-S™ cell lines
• Optimization of feeding strategies will be part of your base medium selection testing in an effort to maximize productivity
• Your CHO cell line is “finicky,” i.e. hard to grow or adapt

Consider CD CHO medium when:
• Your GS CHO cells are recommended to be grown in CD CHO medium
• Your cells have been successfully cultured in CD CHO medium in the past and your process is nearing a regulatory filing (DMF available)

Design Your Own Media Specific to Your Needs
Whether it’s a bench-level experiment or large scale biomanufacturing process, we offer specialized formats, packaging, and quality levels specific to your needs. With Gibco™ cGMP Media Custom Services you can put the manufacturing power and years of technical experience of our R&D and Bioproduction Application Specialists to work for you.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Cell LineCHO DG44
Concentration1 X
Endotoxin LevelLow
FormatBottle(s)
Included AntibioticsNo antibiotics
Product LineGibco
Product TypeCD DG44 Medium
Quantity1 L
Shipping ConditionRoom Temperature
ClassificationAnimal Origin-free, Chemically-defined, Protein-free, Serum-free
Culture TypeSuspension Cell Culture
FormLiquid
Serum LevelSerum-free
With AdditivesSodium Pyruvate, Sodium Bicarbonate, Thymidine, Hypoxanthine
Without AdditivesNo Glutamine, No Phenol Red
Unit SizeEach
Contents & Storage
Store in refrigerator (2–8°C) and protect from light.