Methyl benzoate, 99%
Methyl benzoate, 99%
Methyl benzoate, 99%
Thermo Scientific Chemicals

Methyl benzoate, 99%

CAS: 93-58-3 | C8H8O2 | 136.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
2.5 L
10 L
Packaging:
View all
Glass bottle
Catalog number 126345000
Price (USD)
49.65
Special offer
Online exclusive
Ends: 28-Dec-2026
58.60
Save 8.95
Each
Quantity:
500 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
49.65
Special offer
Online exclusive
Ends: 28-Dec-2026
58.60
Save 8.95
Each
Chemical Identifiers
CAS93-58-3
IUPAC Namemethyl benzoate
Molecular FormulaC8H8O2
InChI KeyQPJVMBTYPHYUOC-UHFFFAOYSA-N
SMILESCOC(=O)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
GC>=98.5 %
Specific gravity(25/25°C) 1.084 to 1.088
Appearance (Form)Liquid
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only