Methyl 5-bromovalerate, 97%
Methyl 5-bromovalerate, 97%
Methyl 5-bromovalerate, 97%
Thermo Scientific Chemicals

Methyl 5-bromovalerate, 97%

CAS: 5454-83-1 | C6H11BrO2 | 195.06 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
25 g
Packaging:
Glass bottle
Catalog number 126460100
Price (USD)
81.65
Special offer
Online exclusive
Ends: 28-Dec-2026
95.40
Save 13.75
Each
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
81.65
Special offer
Online exclusive
Ends: 28-Dec-2026
95.40
Save 13.75
Each
Specifications
Assay Percent Range97%
CAS5454-83-1
Chemical Name or MaterialMethyl 5-bromovalerate
ColorBrown to Colorless
Density1.3600g/mL
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only