3-Methyl-1-butanol, 98%, pure, mixture of Isomers
3-Methyl-1-butanol, 98%, pure, mixture of Isomers
3-Methyl-1-butanol, 98%, pure, mixture of Isomers
Thermo Scientific Chemicals

3-Methyl-1-butanol, 98%, pure, mixture of Isomers

CAS: 123-51-3 | C5H12O | 88.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
1 L
2.5 L
25 L
Packaging:
View all
Glass bottle
Catalog number 126480025
Price (USD)
204.65
Online Exclusive
227.00
Save 22.35 (10%)
Each
Quantity:
2.5 L
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
204.65
Online Exclusive
227.00
Save 22.35 (10%)
Each
Chemical Identifiers
CAS123-51-3
IUPAC Name3-methylbutan-1-ol
Molecular FormulaC5H12O
InChI KeyPHTQWCKDNZKARW-UHFFFAOYSA-N
SMILESCC(C)CCO
View more
SpecificationsSpecification SheetSpecification Sheet
Color scale=<20 APHA
Water=<0.5 % (K.F.)
GC>=97.5 %
Infrared spectrumConforms
Refractive index1.4050 to 1.4090 (20°C, 589 nm)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only