FreeStyle™ CHO Expression Medium, 6 x 1 L - FAQs
FreeStyle™ CHO Expression Medium, 6 x 1 L - FAQs
View additional product information for FreeStyle™ CHO Expression Medium - FAQs (12651022, 12651014)
1 product FAQs found
Can I load the FreeStyle 293 Expression Medium or FreeStyle CHO Expression Medium directly onto a nickel column for His-tagged protein purification?
With FreeStyle 293 Expression Medium, we would recommend harvesting the cells a little earlier to reduce any nonspecific binding by host cell proteins and then filtering the culture supernatant through a 0.2 or 0.1 µM membrane before loading onto the column. On the other hand, FreeStyle CHO Expression Medium contains EDTA that will strip the nickel column. The medium can be subjected to dialysis or buffer exchange prior to loading on the column, or the EDTA can be chelated by adding 1-3 mg/L NiSO4 or NiCl to the supernatant. Iron or cobalt salts can also be used.