4-Methylcatechol, 98%
4-Methylcatechol, 98%
4-Methylcatechol, 98%
Thermo Scientific Chemicals

4-Methylcatechol, 98%

CAS: 452-86-8 | C7H8O2 | 124.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
500 g
Packaging:
Glass bottle
Catalog number 126515000
Price (USD)
-
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS452-86-8
IUPAC Name4-methylbenzene-1,2-diol
Molecular FormulaC7H8O2
InChI KeyZBCATMYQYDCTIZ-UHFFFAOYSA-N
SMILESCC1=CC=C(O)C(O)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to brown
Appearance (Form)Powder and/or chunks
Melting point63°C to 68°C
Infrared spectrumConforms
GC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only